4-[(2,6-Dichlorobenzyl)oxy]benzoic acid

Supplier: Matrix Scientific

040069-500MG
101848-776EA 193.85 USD
101848-776
4-[(2,6-Dichlorobenzyl)oxy]benzoic acid
4-[(2,6-Dichlorobenzyl)oxy]benzoic acid

Hazard: Irritant

Formula: C₁₄H₁₀Cl₂O₃
MDL Number: MFCD09710361
CAS Number: 149289-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR