2-[(3-Methylbenzyl)oxy]benzoic acid

Supplier: Matrix Scientific

040068-500MG
101848-774EA 193.85 USD
101848-774
2-[(3-Methylbenzyl)oxy]benzoic acid
2-[(3-Methylbenzyl)oxy]benzoic acid

Hazard: Irritant

Formula: C₁₅H₁₄O₃
MDL Number: MFCD05267697
CAS Number: 744242-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR