2-[(3-Fluorobenzyl)oxy]-3-methoxybenzoic acid

Supplier: Matrix Scientific

040063-500MG
101848-764EA 193.85 USD
101848-764
2-[(3-Fluorobenzyl)oxy]-3-methoxybenzoic acid
2-[(3-Fluorobenzyl)oxy]-3-methoxybenzoic acid

Hazard: Irritant

Formula: C₁₅H₁₃FO₄
MDL Number: MFCD12028124
CAS Number: 1142201-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR