2-[(4-Methylbenzyl)oxy]benzoic acid

Supplier: Matrix Scientific

040061-500MG
101848-760EA 144.9 USD
101848-760
2-[(4-Methylbenzyl)oxy]benzoic acid
2-[(4-Methylbenzyl)oxy]benzoic acid

Hazard: Irritant

Formula: C₁₅H₁₄O₃
MDL Number: MFCD09719413
CAS Number: 52803-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR