3-Methoxy-4-[(4-methylbenzyl)oxy]benzoic acid
Supplier: Matrix Scientific
040060-500MG
101848-758EA
144.9
USD
101848-758
3-Methoxy-4-[(4-methylbenzyl)oxy]benzoic acid
3-Methoxy-4-[(4-methylbenzyl)oxy]benzoic acid
Hazard: Irritant
Formula:
C₁₆H₁₆O₄ |
MDL Number:
MFCD06634204 CAS Number: 838887-77-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...