3-[(4-Methylbenzyl)oxy]benzoic acid
Supplier: Matrix Scientific
040059-500MG
101848-756EA
193.85
USD
101848-756
3-[(4-Methylbenzyl)oxy]benzoic acid
3-[(4-Methylbenzyl)oxy]benzoic acid
Hazard: Irritant
Formula:
C₁₅H₁₄O₃ |
MDL Number:
MFCD09712906 CAS Number: 307302-08-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...