5-Bromo-2-[(2-chloro-4-fluorobenzyl)oxy]benzoic acid

Supplier: Matrix Scientific

040053-500MG
101848-744EA 193.85 USD
101848-744
5-Bromo-2-[(2-chloro-4-fluorobenzyl)oxy]benzoic acid
5-Bromo-2-[(2-chloro-4-fluorobenzyl)oxy]benzoic acid

Hazard: Irritant

Formula: C₁₄H₉BrClFO₃
MW: 359.59 g/mol
MDL Number: MFCD12028120
CAS Number: 1039813-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR