2-[(2-Chloro-4-fluorobenzyl)oxy]benzoic acid

Supplier: Matrix Scientific

040052-500MG
101848-742EA 193.85 USD
101848-742
2-[(2-Chloro-4-fluorobenzyl)oxy]benzoic acid
2-[(2-Chloro-4-fluorobenzyl)oxy]benzoic acid

Hazard: Irritant

Formula: C₁₄H₁₀ClFO₃
MDL Number: MFCD12028119
CAS Number: 1019348-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR