2-[(4-Fluorobenzyl)oxy]-3-methoxybenzoic acid

Supplier: Matrix Scientific

040051-500MG
101848-740EA 193.85 USD
101848-740
2-[(4-Fluorobenzyl)oxy]-3-methoxybenzoic acid
2-[(4-Fluorobenzyl)oxy]-3-methoxybenzoic acid

Hazard: Irritant

Formula: C₁₅H₁₃FO₄
MDL Number: MFCD04488434
CAS Number: 861433-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR