4-[(2-Methylbenzyl)oxy]benzoic acid

Supplier: Matrix Scientific

040050-500MG
101848-738EA 193.85 USD
101848-738
4-[(2-Methylbenzyl)oxy]benzoic acid
4-[(2-Methylbenzyl)oxy]benzoic acid

Hazard: Irritant

Formula: C₁₅H₁₄O₃
MDL Number: MFCD11529222
CAS Number: 149289-01-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR