5-Bromo-2-[(2-methylbenzyl)oxy]benzoic acid

Supplier: Matrix Scientific

040048-500MG
101848-734EA 193.85 USD
101848-734
5-Bromo-2-[(2-methylbenzyl)oxy]benzoic acid
5-Bromo-2-[(2-methylbenzyl)oxy]benzoic acid

Hazard: Irritant

Formula: C₁₅H₁₃BrO₃
MDL Number: MFCD09707674
CAS Number: 938153-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR