5-Bromo-2-[(4-chlorobenzyl)oxy]benzoic acid

Supplier: Matrix Scientific

040047-500MG
101848-732EA 193.85 USD
101848-732
5-Bromo-2-[(4-chlorobenzyl)oxy]benzoic acid
5-Bromo-2-[(4-chlorobenzyl)oxy]benzoic acid

Hazard: Irritant

Formula: C₁₄H₁₀BrClO₃
MDL Number: MFCD09717777
CAS Number: 62176-36-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR