4-[(4-Chlorobenzyl)oxy]-3-methoxybenzoic acid

Supplier: Matrix Scientific

040046-500MG
101848-730EA 193.85 USD
101848-730
4-[(4-Chlorobenzyl)oxy]-3-methoxybenzoic acid
4-[(4-Chlorobenzyl)oxy]-3-methoxybenzoic acid

Hazard: Irritant

Formula: C₁₅H₁₃ClO₄
MDL Number: MFCD02063085
CAS Number: 113457-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR