3-Bromo-4-isopropoxy-5-methoxybenzoic acid

Supplier: Matrix Scientific

040045-500MG
101848-728EA 144.9 USD
101848-728
3-Bromo-4-isopropoxy-5-methoxybenzoic acid
3-Bromo-4-isopropoxy-5-methoxybenzoic acid

Hazard: Irritant

Formula: C₁₁H₁₃BrO₄
MDL Number: MFCD09722735
CAS Number: 938243-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR