2-Bromo-4-ethoxy-5-methoxybenzoic acid
Supplier: Matrix Scientific
040044-500MG
101848-726EA
165.6
USD
101848-726
2-Bromo-4-ethoxy-5-methoxybenzoic acid
2-Bromo-4-ethoxy-5-methoxybenzoic acid
Hazard: Irritant
Formula:
C₁₀H₁₁BrO₄ |
MDL Number:
MFCD06197003 CAS Number: 838814-43-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...