2-Bromo-5-methoxy-4-propoxybenzoic acid

Supplier: Matrix Scientific

040042-500MG
101848-722EA 144.9 USD
101848-722
2-Bromo-5-methoxy-4-propoxybenzoic acid
2-Bromo-5-methoxy-4-propoxybenzoic acid

Hazard: Irritant

Formula: C₁₁H₁₃BrO₄
MDL Number: MFCD12028116
CAS Number: 1024326-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR