3-Bromo-5-methoxy-4-propoxybenzoic acid

Supplier: Matrix Scientific

040041-500MG
101848-720EA 193.85 USD
101848-720
3-Bromo-5-methoxy-4-propoxybenzoic acid
3-Bromo-5-methoxy-4-propoxybenzoic acid

Hazard: Irritant

Formula: C₁₁H₁₃BrO₄
MDL Number: MFCD03973126
CAS Number: 554407-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR