3-Bromo-4,5-diethoxybenzoic acid

Supplier: Matrix Scientific

040040-500MG
101848-718EA 193.85 USD
101848-718
3-Bromo-4,5-diethoxybenzoic acid
3-Bromo-4,5-diethoxybenzoic acid

Hazard: Irritant

Formula: C₁₁H₁₃BrO₄
MDL Number: MFCD05997652
CAS Number: 712294-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR