2-Bromo-4,5-diethoxybenzoic acid

Supplier: Matrix Scientific

040039-500MG
101848-716EA 193.85 USD
101848-716
2-Bromo-4,5-diethoxybenzoic acid
2-Bromo-4,5-diethoxybenzoic acid

Hazard: Irritant

Formula: C₁₁H₁₃BrO₄
MDL Number: MFCD05992346
CAS Number: 708285-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR