3-Bromo-4-ethoxy-5-methoxybenzoic acid
Supplier: Matrix Scientific
040038-500MG
101848-714EA
193.85
USD
101848-714
3-Bromo-4-ethoxy-5-methoxybenzoic acid
3-Bromo-4-ethoxy-5-methoxybenzoic acid
Hazard: Irritant
Formula:
C₁₀H₁₁BrO₄ |
MDL Number:
MFCD03976236 CAS Number: 671790-48-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...