3-Methoxy-4-[(2-methylbenzyl)oxy]benzoic acid

Supplier: Matrix Scientific

040037-500MG
101848-712EA 193.85 USD
101848-712
3-Methoxy-4-[(2-methylbenzyl)oxy]benzoic acid
3-Methoxy-4-[(2-methylbenzyl)oxy]benzoic acid

Hazard: Irritant

Formula: C₁₆H₁₆O₄
MDL Number: MFCD09707912
CAS Number: 938232-46-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR