3-Methoxy-2-[(2-methylbenzyl)oxy]benzoic acid

Supplier: Matrix Scientific

040036-500MG
101848-710EA 193.85 USD
101848-710
3-Methoxy-2-[(2-methylbenzyl)oxy]benzoic acid
3-Methoxy-2-[(2-methylbenzyl)oxy]benzoic acid

Hazard: Irritant

Formula: C₁₆H₁₆O₄
MDL Number: MFCD09714457
CAS Number: 938247-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR