2-[(2-Methylbenzyl)oxy]benzoic acid

Supplier: Matrix Scientific

040035-500MG
101848-708EA 193.85 USD
101848-708
2-[(2-Methylbenzyl)oxy]benzoic acid
2-[(2-Methylbenzyl)oxy]benzoic acid

Hazard: Irritant

Formula: C₁₅H₁₄O₃
MDL Number: MFCD05262735
CAS Number: 743440-26-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR