5-Bromo-2-[(4-bromobenzyl)oxy]benzoic acid

Supplier: Matrix Scientific

040033-500MG
101848-704EA 193.85 USD
101848-704
5-Bromo-2-[(4-bromobenzyl)oxy]benzoic acid
5-Bromo-2-[(4-bromobenzyl)oxy]benzoic acid

Hazard: Irritant

Formula: C₁₄H₁₀Br₂O₃
MDL Number: MFCD09720756
CAS Number: 938241-69-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR