5-Bromo-2-[(2-chlorobenzyl)oxy]benzoic acid

Supplier: Matrix Scientific

040032-500MG
101848-702EA 144.9 USD
101848-702
5-Bromo-2-[(2-chlorobenzyl)oxy]benzoic acid
5-Bromo-2-[(2-chlorobenzyl)oxy]benzoic acid

Hazard: Irritant

Formula: C₁₄H₁₀BrClO₃
MDL Number: MFCD09711195
CAS Number: 62176-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR