4-[(2-Chlorobenzyl)oxy]-3-methoxybenzoic acid

Supplier: Matrix Scientific

040031-500MG
101848-700EA 144.9 USD
101848-700
4-[(2-Chlorobenzyl)oxy]-3-methoxybenzoic acid
4-[(2-Chlorobenzyl)oxy]-3-methoxybenzoic acid

Hazard: Irritant

Formula: C₁₅H₁₃ClO₄
MDL Number: MFCD02628391
CAS Number: 113457-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR