3-[(3-Chlorobenzyl)oxy]benzoic acid

Supplier: Matrix Scientific

040030-500MG
101848-698EA 193.85 USD
101848-698
3-[(3-Chlorobenzyl)oxy]benzoic acid
3-[(3-Chlorobenzyl)oxy]benzoic acid

Hazard: Irritant

Formula: C₁₄H₁₁ClO₃
MDL Number: MFCD00743039
CAS Number: 302569-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR