4-[(3-Chlorobenzyl)oxy]benzoic acid

Supplier: Matrix Scientific

Synonyms: 4-((3-Chlorobenzyl)oxy)benzoic acid

040029-500MG
101848-696EA 153.85 USD
101848-696
4-[(3-Chlorobenzyl)oxy]benzoic acid
4-[(3-Chlorobenzyl)oxy]benzoic acid

Hazard: Irritant

Formula: C₁₄H₁₁ClO₃
MW: 262.69 g/mol
MDL Number: MFCD04114337
CAS Number: 84403-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR