2-[(4-Fluorobenzyl)oxy]benzoic acid

Supplier: Matrix Scientific

040020-500MG
101848-678EA 221.54 USD
101848-678
2-[(4-Fluorobenzyl)oxy]benzoic acid
2-[(4-Fluorobenzyl)oxy]benzoic acid

Hazard: Irritant

Formula: C₁₄H₁₁FO₃
MDL Number: MFCD02724829
CAS Number: 396-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR