4-[(3-Fluorobenzyl)oxy]-3-methoxybenzoic acid
Supplier: Matrix Scientific
040019-500MG
101848-676EA
193.85
USD
101848-676
4-[(3-Fluorobenzyl)oxy]-3-methoxybenzoic acid
4-[(3-Fluorobenzyl)oxy]-3-methoxybenzoic acid
Hazard: Irritant
Formula:
C₁₅H₁₃FO₄ |
MDL Number:
MFCD08070473 CAS Number: 915918-66-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...