2-[(3-Bromobenzyl)oxy]benzoic acid

Supplier: Matrix Scientific

040016-500MG
101848-670EA 193.85 USD
101848-670
2-[(3-Bromobenzyl)oxy]benzoic acid
2-[(3-Bromobenzyl)oxy]benzoic acid

Hazard: Irritant

Formula: C₁₄H₁₁BrO₃
MDL Number: MFCD05263818
CAS Number: 743453-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR