2-(Benzyloxy)-3-methoxybenzoic acid

Supplier: Matrix Scientific

040012-500MG
101848-662EA 98.46 USD
101848-662
2-(Benzyloxy)-3-methoxybenzoic acid
2-(Benzyloxy)-3-methoxybenzoic acid

Hazard: Irritant

Formula: C₁₅H₁₄O₄
Melting Pt: 90 °C
MDL Number: MFCD04594732
CAS Number: 23806-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR