2-[(2-Fluorobenzyl)oxy]-3-methoxybenzoic acid

Supplier: Matrix Scientific

040011-500MG
101848-660EA 193.85 USD
101848-660
2-[(2-Fluorobenzyl)oxy]-3-methoxybenzoic acid
2-[(2-Fluorobenzyl)oxy]-3-methoxybenzoic acid

Hazard: Irritant

Formula: C₁₅H₁₃FO₄
MDL Number: MFCD09721427
CAS Number: 938380-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR