2-(Benzyloxy)-5-bromobenzoic acid
Supplier: Matrix Scientific
040003-500MG
101848-644EA
193.85
USD
101848-644
2-(Benzyloxy)-5-bromobenzoic acid
2-(Benzyloxy)-5-bromobenzoic acid
Hazard: Irritant
Formula:
C₁₄H₁₁BrO₃ |
MDL Number:
MFCD04612889 CAS Number: 62176-31-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...