4-(Cyclopentyloxy)-3-ethoxybenzoic acid

Supplier: Matrix Scientific

040002-500MG
101848-642EA 193.85 USD
101848-642
4-(Cyclopentyloxy)-3-ethoxybenzoic acid
4-(Cyclopentyloxy)-3-ethoxybenzoic acid

Hazard: Irritant

Formula: C₁₄H₁₈O₄
MDL Number: MFCD11522651
CAS Number: 1019350-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR