3-Ethoxy-4-propoxybenzoic acid

Supplier: Matrix Scientific

040000-500MG
101848-638EA 144.9 USD
101848-638
3-Ethoxy-4-propoxybenzoic acid
3-Ethoxy-4-propoxybenzoic acid

Hazard: Irritant

Formula: C₁₂H₁₆O₄
MDL Number: MFCD04608636
CAS Number: 52009-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR