3-Methoxy-4-(pentyloxy)benzoic acid

Supplier: Matrix Scientific

039998-500MG
101848-634EA 153.85 USD
101848-634
3-Methoxy-4-(pentyloxy)benzoic acid
3-Methoxy-4-(pentyloxy)benzoic acid

Hazard: Irritant

Formula: C₁₃H₁₈O₄
Melting Pt: 134…136 °C
MDL Number: MFCD02726427
CAS Number: 95459-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR