4-Butoxy-3-methoxybenzoic acid
Supplier: Matrix Scientific
039996-500MG
101848-630EA
121.54
USD
101848-630
4-Butoxy-3-methoxybenzoic acid
4-Butoxy-3-methoxybenzoic acid
Hazard: Irritant
Formula:
C₁₂H₁₆O₄ |
MDL Number:
MFCD03949026 CAS Number: 3535-34-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...