4-[(2-Chlorobenzyl)oxy]benzoic acid

Supplier: Matrix Scientific

039995-500MG
101848-628EA 193.85 USD
101848-628
4-[(2-Chlorobenzyl)oxy]benzoic acid
4-[(2-Chlorobenzyl)oxy]benzoic acid

Hazard: Irritant

Formula: C₁₄H₁₁ClO₃
MDL Number: MFCD04554128
CAS Number: 84403-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR