4-Isopropoxy-3-methoxybenzoic acid

Supplier: Matrix Scientific

039994-500MG
101848-626EA 75.38 USD
101848-626
4-Isopropoxy-3-methoxybenzoic acid
4-Isopropoxy-3-methoxybenzoic acid

Hazard: Irritant

Formula: C₁₁H₁₄O₄
MDL Number: MFCD01993664
CAS Number: 3535-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR