3-Methoxy-4-propoxybenzoic acid

Supplier: Matrix Scientific

039993-500MG
101848-624EA 193.85 USD
101848-624
3-Methoxy-4-propoxybenzoic acid
3-Methoxy-4-propoxybenzoic acid

Hazard: Irritant

Formula: C₁₁H₁₄O₄
MDL Number: MFCD03658025
CAS Number: 3535-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR