2-Bromo-5-methoxy-4-propoxybenzonitrile

Supplier: Matrix Scientific

039991-500MG
101848-620EA 217.35 USD
101848-620
2-Bromo-5-methoxy-4-propoxybenzonitrile
2-Bromo-5-methoxy-4-propoxybenzonitrile

Hazard: Irritant

Formula: C₁₁H₁₂BrNO₂
MDL Number: MFCD02256772
CAS Number: 515828-87-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR