2-Bromo-5-ethoxy-4-isopropoxybenzonitrile

Supplier: Matrix Scientific

039990-500MG
101848-618EA 290.77 USD
101848-618
2-Bromo-5-ethoxy-4-isopropoxybenzonitrile
2-Bromo-5-ethoxy-4-isopropoxybenzonitrile

Hazard: Irritant

Formula: C₁₂H₁₄BrNO₂
MDL Number: MFCD02256888
CAS Number: 590355-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR