5-Bromo-2-butoxybenzonitrile

Supplier: Matrix Scientific

039988-500MG
101848-614EA 290.77 USD
101848-614
5-Bromo-2-butoxybenzonitrile
5-Bromo-2-butoxybenzonitrile

Hazard: Irritant

Formula: C₁₁H₁₂BrNO
MDL Number: MFCD02257407
CAS Number: 515845-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR