5-Bromo-2-isopropoxybenzonitrile

Supplier: Matrix Scientific

039985-500MG
101848-608EA 290.77 USD
101848-608
5-Bromo-2-isopropoxybenzonitrile
5-Bromo-2-isopropoxybenzonitrile

Hazard: Irritant

Formula: C₁₀H₁₀BrNO
MW: 240.10 g/mol
MDL Number: MFCD02257441
CAS Number: 515832-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR