5-Bromo-2-ethoxybenzonitrile

Supplier: Matrix Scientific

039984-500MG
101848-606EA 290.77 USD
101848-606
5-Bromo-2-ethoxybenzonitrile
5-Bromo-2-ethoxybenzonitrile

Hazard: Irritant

Formula: C₈H₆BrNO
MDL Number: MFCD02257397
CAS Number: 279263-03-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR