2-Bromo-5-ethoxy-4-methoxybenzonitrile

Supplier: Matrix Scientific

039983-500MG
101848-604EA 217.35 USD
101848-604
2-Bromo-5-ethoxy-4-methoxybenzonitrile
2-Bromo-5-ethoxy-4-methoxybenzonitrile

Hazard: Irritant

Formula: C₁₀H₁₀BrNO₂
MDL Number: MFCD02256845
CAS Number: 515847-20-8
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR