2-Bromo-5-ethoxy-4-isopropoxybenzaldehyde oxime

Supplier: Matrix Scientific

039982-500MG
101848-602EA 290.77 USD
101848-602
2-Bromo-5-ethoxy-4-isopropoxybenzaldehyde oxime
2-Bromo-5-ethoxy-4-isopropoxybenzaldehyde oxime

Hazard: Irritant

Formula: C₁₂H₁₆BrNO₃
MDL Number: MFCD02055219
CAS Number: 494858-86-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR