3-Bromo-5-methoxy-4-propoxybenzonitrile

Supplier: Matrix Scientific

039979-500MG
101848-596EA 217.35 USD
101848-596
3-Bromo-5-methoxy-4-propoxybenzonitrile
3-Bromo-5-methoxy-4-propoxybenzonitrile

Hazard: Irritant

Formula: C₁₁H₁₂BrNO₂
MDL Number: MFCD02256505
CAS Number: 515848-04-1
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR