2-Bromo-4-isopropoxy-5-methoxybenzonitrile

Supplier: Matrix Scientific

039976-500MG
101848-590EA 290.77 USD
101848-590
2-Bromo-4-isopropoxy-5-methoxybenzonitrile
2-Bromo-4-isopropoxy-5-methoxybenzonitrile

Hazard: Irritant

Formula: C₁₁H₁₂BrNO₂
MDL Number: MFCD02256815
CAS Number: 515846-11-4
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR